Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hepcidin (Rat) ELISA Kit, BioVision

Supplier: BioVision
AB285233-96TEST
76313-336EA 1402.46 USD InStock
76313-336
Hepcidin (Rat) ELISA Kit, BioVision
Assays ELISAs
A Sandwich ELISA kit for the qualitative detect Hepcidin in rat serum or plasma.

  • Sensitivity: <9.375 pg/ml, Detection Range: 15.625 to 1000 pg/ml, No significant cross-reactivity or interference was observed
  • Easy, convenient and time-saving method to measure the level of Hepcidin in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Hepcidin is a liver-produced hormone that constitutes the main circulating regulator of iron absorption and distribution across tissues. Acts by promoting endocytosis and degradation of ferroportin, leading to the retention of iron in iron-exporting cells and decreased flow of iron into plasma. Controls the major flows of iron into plasma: absorption of dietary iron in the intestine, recycling of iron by macrophages, which phagocytose old erythrocytes and other cells, and mobilization of stored iron from hepatocytes. BioVision’s Hepcidin ELISA kit is a sandwich ELISA assay for the quantitative measurement of Hepc in rat serum, plasma and cell culture supernatants. The density of color is proportional to the amount of Hepcidin captured from the samples.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty