Would you like to log in again?
Macro Cuvette with PTFE Cover and Magnetic Stirrer Slot (Lightpath: 10 mm), Type 1MS
Supplier: FireFlySci
1MSUV10 1MSG10 1MSIR10
76314-052EA 271.99 USD InStock
76314-052 76314-054 76314-056
Macro Cuvette with PTFE Cover and Magnetic Stirrer Slot (Lightpath: 10 mm), Type 1MS
Cuvettes Spectrophotometer Cuvettes
A Type 1 standard spectrophotometer cuvette that comes with a circular groove, which fits a P73 Magnetic Stirrer. The P73 is agitated by a P82 magnetic cuvette stirrer and the rotating P73 creates a vortex, mixing the solution while not interfering with measurements.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in different materials: Optical Glass, UV Quartz, IR Quartz
  • Available in 10 mm lightpath
  • Two windows optically polished
  • Includes PTFE cover
Features circular channel at the bottom in the cell that is 3mm deep and 7mm wide, which fits a standard P73 macro cuvette stirrer. Includes P73 Stirrer for 10 mm cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty