Would you like to log in again?
MAT STICKY BLUE 30LAYERS/PAD 36X72IN CS1
Supplier: VWR International
VWR Mat Sticky Bl 30Lyr/Pad 36X72In Cs4^ VWR* PureStep Sticky Mat, Color: blue, Dimension: 36x72inch, removes/contains dirt and dust from foot traffic and equipment wheels, Consists of 30 sheets of 1.5mil LDPE film, Each sheet has a 0.3mil thick acrylic-based, pressure-sensitive, size: 30 Layer/pad
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
MAT STICKY BLUE 30LAYERS/PAD 36X72IN CS1 76314-470 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty