Would you like to log in again?
VWR® Horizontal MAXI MR Gel Electrophoresis System
Supplier: VWR
VWR®
81-2025R
76314-682EA 1866.99 USD InStock
76314-682
VWR® Horizontal MAXI MR Gel Electrophoresis System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
VWR® MAXI MR model includes a buffer circulation system, powered by the electrodes themselves.
The hydrogen bubbles produced at the cathode pass up a diagonally positioned tube connected to the two sides of the tank producing a gentle circulation of the buffer. The result is a no pH and ionic gradient - without the need of mechanical pumps.
Delivery information: Includes integral buffer circulation, safety lid with retractable sheathed leads, UVT gel tray with gasketed end gates, three combs- one each 16 well, 24 well and 36 well (1.5 mm thick).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty