Would you like to log in again?
Manual Metal-Handle Utility Knife, Slice®
Supplier: Slice
10490
76314-962CS 244.38 USD InStock
76314-962
Manual Metal-Handle Utility Knife, Slice®
Knives
With the shape of a standard utility knife, the 10490 is sure to appeal to those looking for a safer blade in a familiar handle.
  • Installed blade type: rounded tip
  • Durable metal handle with heavy texture finish
  • Manual retraction with 3 cutting positions
  • Blade garage™ to store one extra blade
  • Ambidextrous design
  • Finger-friendly® blade cuts effectively
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
  • 100% zirconium oxide blade
  • Blade edge lasts up to 11.2 times longer than metal
  • Fewer blade changes result in fewer injuries
  • Compatible blades: 10523, 10524, 10525
  • No-tool blade change
A manual-retract slider offers three fixed cutting positions for different tasks, as well as a fully retracted position for safe storage. Keep spare Slice® blades handy with the onboard blade garage, which is designed to hold one spare blade. Choose from compatible rounded-tip blades (10524), pointed-tip blades (10525), or serrated blades (10523). All Slice® blades feature a finger-friendly® edge to reduce lacerations. They also offer superior wear resistance, as Slice® blades last up to 11 times longer than steel blades. In addition, Slice® blades are chemically inert, which means they never rust. The ambidextrous 10490 ships with a rounded-tip blade.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty