Would you like to log in again?
GLOVES ISOLATOR CGL NS NITRILE SZ12 CS40
Supplier: Ansell Healthcare
Gloves Nitrile 33In/840Mm 12In 9.75 Cs40^ Bioclean* Cleanroom Laundered NITRILER ABS and Isolator Glove, Free from natural rubber latex, TSE or any animal derived material, Color: white, Abrasion, Ambidextrous hand shape, smooth surface, 12 mil thick, Port size: 12in, Length: 33in/840mm, Size: 9.75
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES ISOLATOR CGL NS NITRILE SZ12 CS40 76316-256 CGL36NIT59 Case of 40 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty