Would you like to log in again?
BioClean™ Nitrile Clean Non Sterile RABS/Isolator Gloves, Ansell
Supplier: Ansell
BioClean™
CGL10NIT59 CGL20NIT59 CGL36NIT59 CGL10NIT59
76316-253CS 6673.1 USD InStock
76316-253 76316-255 76316-257 CA76316-255
BioClean™ Nitrile Clean Non Sterile RABS/Isolator Gloves, Ansell
Gloves
Non sterile RABS/Isolator gloves.
  • Tested against ASTM D6978-05 for handling chemo drug
  • 100% water leak tested
  • Ultra-clean surface ensures product protection
  • Autoclave compatible
  • ISO 4 Cleanroom laundered and packaged
  • Resistant to VHP, IPA, and disinfectants
BioClean™ RABS and isolator gloves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production areas, the CGL series of gloves are cleanroom processed and packed, and are available in a range of port sizes.
Certifications: ASTM D6978, CE 0598, EC 1935/2004, EN 388:2016, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 Part 2, Category III.
Packaging: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white correx polyethylene box (20 pieces).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty