Would you like to log in again?
GLOVES ISOLATOR GGL ST 12-14 IN SZ9 CS20
Supplier: Ansell Healthcare
Gloves Nitrile Sterile 36 In 9.75 Cs20^ Bioclean* Gloves, GGL 36in, Material: Nitrile, ISO Class 4 & EU GMP Grade A, 840mm (22in) Nitrile contains no natural latex proteins, Smooth,5 Finger Ambidextrous, 100% air leak tested, Minimize hand fatigue, Color: White, Gamma irradiation, Size: 9.75
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES ISOLATOR GGL ST 12-14 IN SZ9 CS20 76316-260 GGL36NIT59 Case of 20 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty