Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BioClean™ Nitrile Sterile Isolator Mittens, Ansell

Supplier: Ansell
BioClean™
GGL30NITM9
76316-262CS 8775.51 USD InStock
76316-262
BioClean™ Nitrile Sterile Isolator Mittens, Ansell
Gloves
Validated sterile nitrile RABS/Isolator Mitten for 254 mm (10") ports.

  • Tested against ASTM D6978-05 for handling chemo drug
  • 100% water leak teste
  • Ultra-clean surface ensures product protection
  • Specially designed to minimize hand fatigue
  • Fits 254 mm (10")port
  • Autoclave compatible
  • ISO 4 Cleanroom laundered and packaged
  • Resistant to VHP, IPA, and disinfectants |

BioClean™ validated sterile RABS and Isolator Mittens are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production  areas, the GGL mittens are fully validated for sterility with an SAL (Sterility Assurance Level) of 10⁻⁶.

Certifications: ASTM D6978, CE 0598, EC 1935/2004, ISO 11137-1:2006, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 388:2016, EN 455 Part 2, Category III.

Packaging: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white Correx polyethylene box (20 pieces).

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty