Would you like to log in again?
TORQUE TOOL TESTER TTC-E-800 19MM 0.05 S
Supplier: SEALS USA INC
Torque Tool Tester 800 N-M (590 Ft-Lb)^ Torque tool tester, Simple operation test instrument for quickly checking torque tool performance, provide long operation life and power flexibility with the ability to work, Power: 3.6 V dc, Accuracy: +/- 0.3% of reading, Capacity: 800 N-m (590 ft-lb)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TORQUE TOOL TESTER TTC-E-800 19MM 0.05 S 76317-602 TTC-E-800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty