Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

VELP® OHS Digital Overhead Mixer Series, VELP Scientifica

Supplier: VELP Scientifica
F20110494 F20110493 F20110490 F20110492 F20110491
76318-382EA 2216.08 USD InStock
76318-382 76318-372 76318-374 76318-378 76318-368
VELP® OHS Digital Overhead Mixer Series, VELP Scientifica
Stirrers Overhead Stirrers
VELP® OHS Series overhead stirrers are engineered to guarantee state-of-the-art stirring power maintaining the maximum torque at any speed and to resist heavy duty for years and encompass the latest technology at a great value.

  • 230 V model
  • Digital display with torque indicators
  • Integrated timer
  • Smart chuck technology for 1 to 13 mm shafts
  • Key lock safety feature
  • Double speed gear (on 200 N*cm model only)
  • Brushless motors: 30 to 2000 rpm, ±1 rpm

Safe Operations - Extremely resistant sealed case. IP 54 Protection class is a guarantee of durability also when working in aggressive environment. Full protection in case of overload, overtemperature and over current. Lock key to prevent unintentional modification of the settings. Easy knob to navigate the menu and to activate the stirring. OHS Digital are designed for the longest lifespan and guaranteed by 3 years warranty.

High Performance — Digital display to precisely set the working conditions. Brushless motor ensures stable torque of up to 200 Ncm through the whole speed range. OHS 200 Digital has two Speed gears for maximum flexibility: high-torque or high-speed operation. Electronic speed control fo up to 2000 rpm. Maximum volume of up to 100 liters. Maximum viscosity of up to 100.000 mPa. SpeedServo™ torque compensation technology. Silent motor even at the maximum speed.

Premium Features – The new SmartChuck™ makes easy and convenient the shaft changing without any tool. Integrated timer for unattended operation with automatic switch off. Torque monitoring display with four LED segments for real-time information on viscosity change. USB interface for installing Software updates.

Certifications: Meets IP54 housing protection rating. cNemko certified.

Ordering information: Supplied without stand and accessories, optional accessories available separately.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty