Would you like to log in again?
pH Probe Holder and Ferrules, Ace Glass
Supplier: Ace Glass
5277-12 5277-02
76318-480PK 68.97 USD InStock
76318-480 76214-390
pH Probe Holder and Ferrules, Ace Glass
Electrode Holders
pH probe holder for use with Ace Glass Scale-Up Series benchtop reactor systems from 2000 to 6000 ml.
  • Designed specifically for Cole-Parmer refillable general purpose probe
Unwetted section: 115L×16D mm
Wetted section: 12.3D mm
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty