Would you like to log in again?
Photochemical Turntable Reactor, Ace Glass Incorporated
Supplier: Ace Glass
7891-30 7891-35
89061-404EA 12416.4 USD InStock
89061-404 76318-574
Photochemical Turntable Reactor, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
ACE photochemical reactor with sample roundtable feature for the determination of relative and absolute quantum yields
Polished and Colored Filter Glasses are a combination of either Corning Glass Filter cs 0-52 and Corning Glass Filter cs 7-37 or Corning Glass Filter cs 7-60 isolates 3650 line. 51 x 51 mm. For use in four-sided filter chamber supplied with the Photochemical turntable reactor.
When Corning Glass Filter cs 7-54 is used in conjunction with a filter solution of potassium chromate and sodium hydroxide less than 3% of the 3340 line passes. The solution may be circulated via pump as a coolant through well. 51x51 mm. or use in four-sided filter chamber supplied with the Photochemical turntable reactor.
Features adjustable height slots for up to 33 sample tubes in outer circle and 18 tubes in inner circle, 13 mm O.D. Samples rotate equidistantly around lamp at 6 rpm. Chamber between lamp and samples holds four, 51 mm square, removable flat filters. Unit is constructed of anodized aluminum, brass and PTFE. Measures 25.4 cm diameter at base × 61 cm high at top of motor stand. Except for motor, reactor is completely immersible and can be easily disassembled for cleaning.
Ordering information: Available in 115 V, 50/60 Hz. or 230 V, 50 Hz. Supplied with two-wire, ground NEMA plug. Complete unit includes motor, but does not include filters, immersion well, lamp, power supply or sample tubes.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty