Would you like to log in again?
Ferrule Gripper Sets, Ace Glass Incorporated
Supplier: Ace Glass
12729-05 12729-03 12729-06 12729-17 12729-18 12729-04 12729-12 12729-15 12729-07 12729-08 12729-13 12729-01 12729-09
76318-540EA 30.1 USD InStock
76318-540 76318-538 76318-604 76318-616 76318-606 76318-608 76318-598 76318-610 76318-600 76318-612 76318-602 76318-536 76318-614
Ferrule Gripper Sets, Ace Glass Incorporated
Fittings and Connectors
Replacement ferrule/gripper sets for ACE PTFE tube compression fittings.
Delivery information: Each set consists of one each PTFE ferrule and PVDF gripper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty