Would you like to log in again?
ARM-D Kit, ampC (RUO), Streck
Supplier: Streck
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
250044
76318-694EA 1855.59 USD InStock
76318-694
ARM-D Kit, ampC (RUO), Streck
Assays qPCR Assays
The Streck ARM-D Kit, AmpC, is a comprehensive, TaqMan based method for the detection of plasmid based antibiotic resistance gene families in Gram Negative Bacteria.
  • Dectects the six major ampC resistance families
  • The only commercially available multiplex kit to identify plasmid-mediated AmpCs
  • Understand resistance mechanisms prior to obtaining susceptibility results
  • Works on a large variety of instrumentation
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty