Would you like to log in again?
VWR COVERALL BASICPROTECTION BK 2XL CS25
Supplier: VWR International
VWR Coverall Basic Protection Bk 2Xl CS25^ VWR Basic Protection Coverall Made Of 50Gsm Black Sms, Elastic Wrist, Ankle And Back, Zip Down Front, 2Xl Size, Serged Seams Case Of 25
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR COVERALL BASICPROTECTION BK 2XL CS25 76318-836 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty