Would you like to log in again?
GLOVES NITRILE LONG CUFF SZ11 PK1 12PAIR
Supplier: Ansell Healthcare
Glove Nitrile Long Cuff Sz11 PK1 12PAIR^ AlphaTec 11.8* Nitrile Glove, Light Duty Chemical/Liquid/Splash, Straight Long Cuff,Lozenge Grip,AQL.65,Silicone Free, Latex Free, 8mil thickness allows dexterity and tactility without a loss of durability, NBR offering protection against acids, Size: 11
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES NITRILE LONG CUFF SZ11 PK1 12PAIR 76319-010 37310110 Case of 12 (1/ Itm/Bx) Retrieving
Pack of 1 (1/Itm/Bx) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty