Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ergolux Jr. Lab Stools

Supplier: BevCo Precision Manufacturing
Ergolux Jr.
J3000-GRP J3000-BLK J3350-BLK J3300-GRP J3050-GRP J3350-GRP J3300-BLK J3050-BLK
76319-124EA 358.14 USD InStock
76319-124 76319-122 76319-100 76319-120 76319-098 76319-096 76319-094 76319-092
Ergolux Jr. Lab Stools
Chairs
Advanced stress relief With the Ergolux Jr. series.

  • The 2.5" thick ultrasoft, high-density, visco-elastic memory foam seat provides resilient, long-lasting ergonomic support
  • The 14.5" diameter seat is covered by puncture, water, and stain-resistant polyurethane
  • Stool is non-slip and easy-to-clean
  • Easy-to-use pneumatic height adjustment
  • Sturdy 25" diameter black nylon base
  • Dual-wheel hard floor casters
  • Black seat color
  • 300 lb. weight capacity
  • For standard environments

The stress-relieving qualities of the stool are heightened because they respond to your body's unique shape, weight, and temperature. The Ergolux Jr. series is ideal for a wide range of applications, including general office, medical, laboratory, and higher education applications.

Certifications: ANSI/BIFMA; GreenGuard Gold.

Ordering information: 12-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty