Would you like to log in again?
LaboPlast® and SteriPlast® Compact Jerrycans, Electroconductive, Bürkle
Supplier: Bürkle
0435-2055 0435-2105 0435-1105 0435-0015 0435-0005 0435-1055
76319-722EA 111.75 USD InStock
76319-722 76319-732 76319-730 76319-728 76319-726 76319-724
LaboPlast® and SteriPlast® Compact Jerrycans, Electroconductive, Bürkle
Bottles Jerricans
These cans suited for use in explosion-proof areas. With a specific surface resistance of <105 ohms, the canister has a very high electrical conductivity. When correctly earthed, it prevents the danger of ignition due to an accumulation of electrostatic charge. The canister thus helps to meet the requirements of the ATEX 137, TRBS2153, and CENELEC 50404 guidelines.
  • HDPE, black, anti-static
  • Electroconductive
  • Available with or without thread support (³/₄")
Plastic industrial canisters are non-rusting and can be disposed of in the recycling system. They can be used for dispensing and storing combustible and highly flammable liquids of explosion groups IIA, IIB, and IIC, such as laboratory waste in industrial applications or mining. Due to the narrow width of 6.5 or 12.5 cm, the space-saving canister requires very little room, making it suitable for use in confined spaces. The ribbed structure and thick sides make it very stable and also allow for flat storage. The handle ensures secure handling and easy transport. For the version with ³/₄" thread support, a stopcock can be mounted, for example the StopCock PA, which is electrically conductive.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty