Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PTFE Barrel Pumps, Ultrapure, Burkle

Supplier: Bürkle
5606-1000 5606-1001 5606-0401 5606-0601 5606-0600 5606-0400
10217-248EA 1140.79 USD InStock
10217-248 76319-734 10217-246 10217-252 10217-250 76319-736
PTFE Barrel Pumps, Ultrapure, Burkle
Pumps Drum/Transfer Pumps
Due to the high chemical resistance of the medium-contacting parts made of PTFE and FEP, even liquids which attack conventional materials such as PP or PVC can now be safely transferred (except for elemental fluorine) with this barrel pump. The barrel pump is also suitable for transferring ultra-pure liquids. The stopcock prevents hazardous vapors from escaping, and stops dripping after transfer.

  • PTFE, white
  • Excellent chemical resistance
  • Immersion tube Ø32mm (1.26")
  • All valves are made of solid PTFE/PVDF
  • Infinitely variable adjustment of pump immersion depth guaranteeing good residual extraction
  • Barrel screw joints for commercially available containers available as accessories

Barrel pump with flexible discharge hose made of PTFE, length of 1.2m (3.94') and stopcock made of PVDF.

Barrel pump with rigid discharge tube with stopcock made of PTFE.

Ordering information: For available accessories, contact your Avantor representative.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty