Would you like to log in again?
Sampling Bags, SteriBag Cleanroom, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SteriBag
5344-8010 5344-8012
76319-744PK 540.83 USD InStock
76319-744 76319-742
Sampling Bags, SteriBag Cleanroom, Bürkle
Bags Sample Bags
The sampling bag SteriBag Cleanroom with double packaging can be placed directly in the clean room. 25 sample bags are placed in double secondary packaging. They can be removed one after the other and are therefore ideally suited for introducing into the clean room. Then the sterile bags can be used directly in the clean room.
  • Sterile, with double packaging
  • RNase, DNase and pyrogen-free
  • PE, transparent
  • Liquid-tight
  • With labelling field
SteriBag Cleanroom is sterile inside and outside. This guarantees contamination-free storage and transport of solid, semi-solid and liquid media. The sample bag is perfect for use in chemical and pharmaceutical applications. The disposable sample bag has a labelling field, saves space and is also easy to transport. The large bag opening makes it easy to pour in the sample.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty