Would you like to log in again?
Palette Knife Spatulas, Stainless Steel, Bürkle
Supplier: Bürkle
5386-0010 5386-0012
76319-750EA 21.38 USD InStock
76319-750 76319-748
Palette Knife Spatulas, Stainless Steel, Bürkle
Spatulas
Similar to a pallet or dispensing chemist's spatula, this palette knife spatula is suitable for applying or distributing viscous media such as pastes and creams, or for spreading them smoothly. That makes cleaning easier and prevents product residue and cleaning agents from accumulating.
  • Stainless steel AISI 304 (1.4301)
  • Very sturdy
  • Manufactured with no edges or crevices
The spatula has a long, narrow, high-gloss polished blade that facilitates precise work. The sturdy handle fits comfortably in the hand. The laboratory spatula is suitable for use in chemical and pharmaceutical applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty