Would you like to log in again?
Forceps, Stainless Steel, Blunt, Straight, Bürkle
Supplier: Bürkle
5386-0304 5386-0300 5386-0302
76319-852EA 9.25 USD InStock
76319-852 76319-850 76319-780
Forceps, Stainless Steel, Blunt, Straight, Bürkle
Forceps
These forceps are suitable for precise laboratory tasks.
  • Stainless steel AISI 304 (1.4301)
  • Flutings at the ends and grip for secure handling
These forceps have a straight shape with blunt ends that are fluted on the inside. This makes it possible to pick up different materials, tissues, or samples accurately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty