Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Biox ID-Color Hinged Freezer Boxes
Supplier: TradeWinds (TWD)
Biox
BX90-8200 BX90-8281
76319-882CS 120.1 USD InStock
76319-882 76319-880
Biox ID-Color Hinged Freezer Boxes
Boxes Cryoboxes
These water-resistant hinged freezer boxes feature a durable plastic-coated cardboard hinge and polypropylene dividers.
  • Coated cardboard
  • 81 and 100 wells
  • PP dividers
  • Alphanumeric coding on lid, sides and bottom
  • Designed for 1.5 and 2.0 ml cryovials
CryoKing 2", water-resistant ID-Color™ cardboard freezer boxes with hinged lid and divider made of PP material. Boxes have marked alphanumeric grid for identification purposes. Boxes come in assorted colors (red, blue, green, yellow and white).
Packaging: 5 Boxes/Sleeve, 4 Sleeves/Case
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty