Would you like to log in again?
Biox Pipette Stand
Supplier: TradeWinds (TWD)
Biox
BX01-2306
76319-892EA 75.92 USD InStock
76319-892
Biox Pipette Stand
Pipette Stands
The pipette stand holds up to 6 single channel pipettes or 1 multi-channel pipette in a linear position.
  • Holds six pipettes
  • Horizontal storage
The universal pipette stand is sturdy and ideal for benchtop storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty