Would you like to log in again?
Sampling Scoops, PS
Supplier: Bürkle
5378-9908 5378-9910 5378-9907 5378-9906
76319-870PK 286.32 USD InStock
76319-870 76319-904 76319-902 76319-868
Sampling Scoops, PS
Scoops
Scoops are ideal for the sampling of powder, granules and pastes.
  • Individually packed, gamma radiation sterilized
Special handle construction and the slightly angled edge stands horizontal and can be laid down without the material trickling out.
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty