Would you like to log in again?
CHILLER HEICHILL3000 550X650X970MM 3000W
Supplier: Heidolph NA, LLC
Chiller Heichill3000 550X650X970Mm 3000W^ Chiller HEI-CHILL 3000, compact design fits effortlessly on a laboratory bench, Cooling capacity at +20 deg C: 3000 W, Temp range: -20 - +40 degC, Pump pressure max: 2 bar, Pump flow max: 20 l/min, Thermal fluid
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHILLER HEICHILL3000 550X650X970MM 3000W 76319-906 036306521 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty