Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

TSX Series High-Performance Lab Refrigerators, Automatic Defrost, Thermo Scientific

Supplier: Thermo Scientific
TSX5005SA TSX2305GD TSX3005SV TSX3005GD TSX1205GA TSX2305GV TSX1205SA TSX3005GA TSX5005GD TSX1205SD TSX5005GA TSX1205SV TSX3005SD TSX3005GV TSX4505GD TSX2305SA TSX2305GA TSX2305SD TSX5005SD TSX1205GV TSX4505GA TSX2305SV TSX3005SA TSX1205GD
76208-592EA 14326.26 USD InStock
76208-592 76320-216 76320-268 75839-106 76320-212 76320-256 76320-258 76208-590 76320-214 75839-102 76320-264 75839-114 76320-266 75839-046 76320-260 76320-292 76320-262 75839-042 76320-294 75839-094 76208-612 76208-586 76208-588 76208-610
TSX Series High-Performance Lab Refrigerators, Automatic Defrost, Thermo Scientific
Refrigerators
Thermo Scientific™ TSX Series high-performance laboratory refrigerators offer a sustainable choice for clinical and laboratory refrigeration.

  • Designed to safely store medical and laboratory storage
  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology
  • Up to 40% less energy usage than conventional refrigerant models
  • Choice of glass or solid door models
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle

Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3 to 7 °C

Designed to safely store medical and laboratory storage, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2°C to 8°C. Units are whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection. These refrigerators offer up to 40% less energy usage than conventional refrigerant models.

Refrigerators feature an automatic defrost and a microprocessor display.

Manufactured in an award-winning, zero-waste to landfill facility. Operating temperature range of 2 to 8 °C, factory setpoint at 5 °C.

5 year warranty on parts and labor, 10 year warranty on compressor parts.

Certifications: All refrigerators are UL, cUL certified. Refrigerators with Europran plug are CE, UL, cUL certified. Refrigerators use natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP).

Accessories information: Optional chart recorders available.

Caution: Note: Dimensions shown below are W×D×H.

Order Now
 

ORDER

SPECIFICATIONS

SERVICES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty