Would you like to log in again?
Axygen® MultiRack Tipstations, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
MR-200-TS-S MR-10XT-TS MR-10XT-TS-S MR-200-TS
76320-712CS 451.34 USD InStock
76320-712 76320-710 76320-708 76320-706
Axygen® MultiRack Tipstations, Corning
Pipette Tips
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack tipstation is packaged in a format with 192 tips per layer and five layers total and can be used with 8 or 12 channel pipettors. This packaging style provides a convenient solution for lab applications such as ELISA, PCR, and NGS library preparation, where thousands of tips are consumed. Axygen® MultiRack tipstation is available with 10 and 200 µl, sterile or non sterile.
Ordering information: Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips and reload configurations.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty