Would you like to log in again?
Axygen® Multirack Bulk Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
MR-200 MR-10XT MR-1000XT
76320-674CS 391.9 USD InStock
76320-674 76320-720 76320-672
Axygen® Multirack Bulk Tips, Corning
Pipette Tips
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
Ordering information: Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty