Would you like to log in again?
GLOVES MICROFLEX GREEN SMALL 6.5-7 PK50
Supplier: Ansell Healthcare
Gloves 93-283 Green Ntrl S 6.5-7.0 PK50^ MICROFLEX* 93-287, 8 mil, Extended Cuff, 11.8in Length, Green, Nitrile, Disposable, Mega Texture grip surface provides a secure grip when h/ling dry, wet, oily or fatty foods / objects. Generous fit for use with underglove, Food Compliant, Size S (6.5-7)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES MICROFLEX GREEN SMALL 6.5-7 PK50 76320-782 93287070 Case of 500 Retrieving
Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty