Would you like to log in again?
GLOVES MICROFLEX GREEN XXL 10.5-11 PK44
Supplier: Ansell Healthcare
Gloves 93-287 Green Xxl 10.5-11.0 PK44^ MICROFLEX* 93-287 Gloves, 8 mil, Extended Cuff, 11.8in, Length, Green, Nitrile, Disposable. Mega Texture grip surface provides a secure grip when handling dry, wet, oily or fatty foods and objects. Generous fit for use with an underglove, Size XXL (10,5-11,0)
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES MICROFLEX GREEN XXL 10.5-11 PK44 76320-798 93287110 Case of 440 Retrieving
Pack of 44 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty