Would you like to log in again?
THERMOMETER CALIB -50/300DEG C 0C 100C
Supplier: Bel-Art Products, a Part of SP
Thermometer 0/100C General Calibration^ H-B Frio Temp Calibrated Electronic Verification Thermometer; Range: -50/300C(-58/572F), Features: Alarm, Timer, Probe Length/Cable Length: 105mm Angled Stainless Steel / 1m Stainless Steel, Calibration Points: 0, 100C, Dimensions (W X H): 52 x 18mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
THERMOMETER CALIB -50/300DEG C 0C 100C 76320-924 B61000-1200 Case of 10 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty