Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hei-VAP Core Rotary Evaporators, Heidolph®

Supplier: Heidolph
036040056 036040054 036040060 036040062 036040053 036040066 036040065 036040059 036040065 036040060 036040050 036040055 036040068 036040051 036040061 036040068 036040055 036040067 036040057 036040064 036040066 036040058 036040058 036040050 036040063 036040056 036040051 036040053 036040052 036040061 036040063 036040069
76320-932EA 9057.94 USD InStock
76320-932 76320-810 76320-766 76320-934 76320-802 76320-768 76320-812 76320-774 76320-762 76320-930 76320-776 76320-764 76320-770 76320-772 76320-808 76320-928 76320-804 76320-914 76320-814 76320-806
Hei-VAP Core Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Designed to access the basic parameters rotation speed and heating bath temperature at any time.

  • Clear digital display showing actual and set values
  • Accidental changing of the values can be prevented by the lock function
  • Standby button stops all functions
  • Standby function and residual heat warning at heating bath temperatures above 50 °C
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Overheat protection bath cut-off at 5 °C over set temperature via separate Pt1000
  • Secondary over temperature cut-off 250 °C

Available in Hand Lift (HL) and Motor Lift (ML) Configurations. Large 3.5" LCD display that indicates rotation speed, bath temperature, process timer, and vapor temperature using the Hei-GUIDE control knob to navigate the parameters.

Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values

Certifications: DIN EN 60529 IP 20.

Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty