Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Lucetta® 2 Luminometer

Supplier: Lonza
AAL-1002
76321-328EA 8827 USD InStock
76321-328
Lucetta® 2 Luminometer
Microplate Readers
The Lucetta® 2 luminometer is a single tube luminometer developed for the detection of bioluminescence and chemiluminescence. It can be used as a portable battery-operated instrument as well as with an external power supply and is suitable for detecting all luminescence.

  • Small, portable tube luminometer
  • Stand-alone instrument without PC requirements
  • High sensitivity and linearity
  • Specific measurement mode for running MycoAlert® Mycoplasma Detection Assays
  • Quick single read mode for unprocessed luminescence readings
  • High data storage capacity with maximum storage of up to 2000 single sample readings

Designed with a specific mode for using Lonza’s MycoAlert® Mycoplasma detection assays, it immediately provides assay results as CLEAN, BORDERLINE or CONTAMINATED sample.

The Lucetta® 2 Luminometer can also be used for many other luminescence-based assays, such as cell proliferation and cytotoxicity assays or luciferase reporter gene assays. For these assays, the instrument's single read mode applies.

Technical specifications:
Dimensions:150×280×170 mm (5.9×11.02×6.69")
Weight:Approx. 2 kg (4.4 lbs.)
Power supply input:100 to 240 V AC, 50 to 60 Hz
Output: 5.9 to 6 V DC/2.1 A
Power consumption (instrument):Max. 15 W
Operating temperature and humidity: 15 to 30 °C 80% humidity at 31 °C degreasing linearly to 50% at 40 °C, no condensation

Applications:
MycoAlert® Mycoplasma Detection Assays.
ATP assays, e.g. ViaLight® Cell Proliferation and Cytotoxicity Assay.
Adenylate Kinase assays, e.g. ToxiLight® Non-destructive Cytotoxicity Assay.
Luciferase reporter gene analysis.

Delivery information: Content: 1 Lucetta® 2 luminometer instrument.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty