Would you like to log in again?
VWR® Syringe Pipette Tips, Evolution
Supplier: Avantor
VWR®
ES40017-0017V ES40017-0016V ES40017-0014V ES40017-0015V ES40017-0004V ES40017-0000V ES40017-0005V ES40017-0013V ES40017-0003V ES40017-0001V ES40017-0007V ES40017-0012V ES40017-0006V ES40017-0011V
76321-816PK 194.79 USD InStock
76321-816 76321-826 76321-824 76321-818 76321-588 76321-576 76321-586 76321-574 76321-822 76321-820 76321-578 76321-580 76321-584 76321-582
VWR® Syringe Pipette Tips, Evolution
Pipette Tips
Evolution syringe tips are compatible with most repeating pipettors on the market. Positive displacement provides precise dosing at even the smallest volumes.
  • Syringe tips come with a color coded adapter for quick identification of the correct size tip
  • Seven tip sizes dispense volumes from 1 µl to 10 ml
  • Available sterile and non sterile
All sizes of the Evolution syringe tips provide reliable and repeatable dosing for a wide range of liquids including viscous, volatile, and infectious material. Sterile tips are individually wrapped and certified to be pyrogen, ATP, RNase, and DNase free. Color coded adapters allow for quick identification of the correct size tip. Manufactured from virgin grade polypropylene.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty