Would you like to log in again?
SafeGard® Disposable Protective Deluxe Coveralls with Attached Hood and Elastic Wrists/Ankles, Zipper Closure, Lakeland Industries
Supplier: Lakeland Industries
C8428B-XL C8428B-LG C8428B-2X C8428B-4X C8428B-3X C8428B-5X C8428B-SM C8428B-MD C8428-SM
76322-100CS 368.86 USD InStock
76322-100 76322-102 76322-104 76322-090 76322-092 76322-094 76322-096 76322-074 76322-098
SafeGard® Disposable Protective Deluxe Coveralls with Attached Hood and Elastic Wrists/Ankles, Zipper Closure, Lakeland Industries
Disposable Garments
Disposable protective coveralls made from three lightweight layers of Spunbond-Meltblown-Spunbond (SMS) polypropylene fabric for protection from dirt, grease, grime, and aerosol mist.
  • Spunbond-Meltblown-Spunbond (SMS) polypropylene fabric
  • Deluxe coverall with elastic back and two pockets
  • Serged seams
  • Breathable protection from dry particulates
  • Effective against aerosol mists and light liquid splash
  • Available in white or blue
SafeGard® SMS coveralls are designed to offer protection from dirt, dust, grease, and grime. The Deluxe pattern coverall features an elastic back along with one chest pocket and one hip pocket for improved fit and convenience. The breathable fabric is effective against most dry particulates, aerosols, and light liquid splashes. These coveralls feature an attached hood with elastic wrist and ankle closures for additional protection. A zipper closure allows for easy donning and doffing.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty