Would you like to log in again?
Whatman™ Grade 598 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10312287 10312209 10312287 10312000 10312000 10312209
10034-684PK 24.56 USD InStock
10034-684 CA74330-470 76322-126 CA76322-126 CA11012-762 74330-470
Whatman™ Grade 598 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A thick filter paper with high loading capacity. Combines medium retention with medium-fast to fast filtration speed.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty