VWR® Universal Pipette Tips
Supplier: VWR
VWR®
63305016 63305039 63305017 63305038 63305370 63305029 63305369 63305032 63305046 63305025 63305022 63305041 63305034 63305054 63305374 63305024 63305036 63305027 63305037 63305048 63305372 63305030 63305035 63305371 63305026 63305368 63305020 63305033 63305042 63305018 63305028 63305023 63305040 63305049 63305045 63305019 63305051 63305373 63305047 63305021 63305031 63305050
76322-134CS 801.84 USD InStock
76322-134 76322-156 76322-136 76322-512 76322-138 76322-514 76322-150 76322-152 76322-132 76322-154 76323-402 76322-526 76322-528 76323-400 76323-388 76595-800 76595-788 76322-144 76322-146 76322-520 76322-522 76322-148 76322-524 76323-404 76322-140 76322-142 76323-394 76595-790 76323-392 76595-792 76323-390 76595-794 76595-796 76322-516 76595-798 76322-518 76323-456 76323-454 76323-398 76327-214 76323-452 76323-396
VWR® Universal Pipette Tips
Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with the widest range of pipettes
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty