Would you like to log in again?
SP Wilmad-LabGlass ProCulture® Fernbach Shaker Flasks, SP Industries
Supplier: SP Industries
ProCulture®
BP-3200-098 BP-3225-100 BP-3250-100 BP-3200-100
76323-670EA 198.34 USD InStock
76323-670 76323-588 76323-586 76323-672
SP Wilmad-LabGlass ProCulture® Fernbach Shaker Flasks, SP Industries
Flasks Erlenmeyer Flasks
Commonly used in serum production and ideal for use in organism cultures that require a large surface area to volume ratio, these flasks are available with a beaded rim, GL 45 screw thread, or Delong style neck for accommodating various style stoppers and plugs.
  • Delong style neck reduces splashing to the cap, when on shaker
  • Beaded rim style provides increased durability to the flask
  • Baffles, spaced at 120°, allow for increased oxygen transfer to the Culture medium when used in conjunction with a shaker
  • Autoclavable
Type 1, Class A borosilicate glass construction. Environmentally sustainable when compared to plastic.
Caution: Do not place on direct heat source
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty