Would you like to log in again?
SP Wilmad-LabGlass ProCulture® Delong Style Shaker Flasks, SP Industries
Supplier: SP Industries
ProCulture®
BP-3350-102 BP-3160-312 BP-3160-306 BP-3300-108 BP-3350-112 BP-3160-310 BP-3300-112 BP-3300-102 BP-3350-104 BP-3160-314 BP-3350-110 BP-3350-108 BP-3300-104 BP-3300-106 BP-3300-110 BP-3350-116
76323-668EA 135.68 USD InStock
76323-668 76323-692 76323-680 76323-690 76323-678 76323-666 76323-688 76323-676 76323-686 76323-664 76323-674 76323-696 76323-662 76323-684 76323-694 76323-682
SP Wilmad-LabGlass ProCulture® Delong Style Shaker Flasks, SP Industries
Flasks Erlenmeyer Flasks
Ideal for growing and mixing of cell cultures, these Delong style shaker flasks have long necks to reduce splashing that could reach the closure.
  • Type 1, Class A borosilicate glass construction
  • Designed for polypropylene and stainless steel closures
  • Polypropylene caps have ribbed interior for improved sealing
  • Autoclavable
Baffles, spaced at 120 °C, allow for increased oxygen transfer to the culture medium when used in conjunction with a shaker.
Environmentally sustainable when compared to plastic.
Caution: Do not place on direct heat source.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty