Would you like to log in again?
VWR COVER PIPE/DRUM WHT SMS 20IN CS100
Supplier: VWR International
Vwr Cover Pipe/Drum Wht Sms 20In CS100^ VWR* Cover, designed for large pipes, drums or items approimately 20 inches in size diameter. Made with white SMS 3 layer material. Sewn 1/4 inch latex free braided elastic for a secure fit.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR COVER PIPE/DRUM WHT SMS 20IN CS100 76323-722 Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty