Would you like to log in again?
HELMET LEATHER WELDING WH20 860P RED
Supplier: ORS NASCO, INC.
Helmet Leather Welding Wh20 860P Red^ Leather Welding Helmet, WH20 860P, style: 860P, Lens Coating/Shade: Green; no. 10, color: black, Material: Leather, Dimensions: 13 in inx10 1/4 inx6 in, Window Width x Length: 2 in x 4 1/4 in, Suspension: 117A, Mounting: Lift Front, Resistance: Heat
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HELMET LEATHER WELDING WH20 860P RED 76324-050 138-14531 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty