Would you like to log in again?
HELMET FIBER SHELL WELDING WH20 490P BK
Supplier: ORS NASCO, INC.
Helmet Fiber Shell Welding Wh20 490P Bk^ Fiber Shell Welding Helmet, WH20 490P, style: 490P, Lens Coating/Shade: Green; no. 10, color: black, Material: Vulcanized Fiber, Dimensions: 13 1/2 inx10 1/4 inx8 1/2 in, Window Width x Length: 2 in x 4 1/4 in, Suspension: 117A, Mounting: Fixed Front
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HELMET FIBER SHELL WELDING WH20 490P BK 76324-052 138-14532 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty