Would you like to log in again?
HOLDER WELDING ELECTRODE MANUAL-ARC 300A
Supplier: ORS NASCO, INC.
Holder Welding Electrode Manual-Arc 300A^ Manual-Arc Welding Electrode Holder, Amps: 300 A, Body Material: Copper Alloy, Cable Cap. 2: 4 AWG - 2/0 AWG, Electrode Capacity: 1/4 in, Welding Amp at Duty Cycle: 300 A at 50 %, Fully insulated spring, Integral grooves, Integrated jaw pattern
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOLDER WELDING ELECTRODE MANUAL-ARC 300A 76324-064 138-14680 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty