Would you like to log in again?
PLATE MAG 2 DIOPTER PC 4 1/4 INX2 IN
Supplier: ORS NASCO, INC.
Plate Mag 2 Diopter Pc 4 1/4 Inx2 In^ Mag Plate, 2 Diopter, Rectangular, Material: Polycarbonate, Dimensions: 4 1/4 inx2 in, While providing close vision without glasses, the mag plates are also adaptable to all EQC Filters, Select focal power by comparing prescription lenses
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE MAG 2 DIOPTER PC 4 1/4 INX2 IN 76324-090 138-16063 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty