Would you like to log in again?
FILTER W60 NEXGEN AUTO-DARK GRD9-12
Supplier: ORS NASCO, INC.
Vwr Filter W60 Nexgen Auto-Dark Grd9-12^ VWR W60 NEXGEN Digital Auto-Darkening Filters, Shade 9-12, Digital controls feature variable shade, sensitivity and delay adjustments, Versatile 3-N-1 built-in function: Weld, Torch, Grind, Built-in mag lens holder, Dimension: 3.8 x 2.35in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER W60 NEXGEN AUTO-DARK GRD9-12 76324-100 138-16622 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty