Would you like to log in again?
HEADGEAR HDG20 FACE SHIELD 8X12X0.04 IN
Supplier: ORS NASCO, INC.
Headgear Hdg20 Face Shield 8X12X0.04 In^ Face Shield Headgear, HDG20, model: 131-B, color: clear, Suspension: Ratchet, Dimensions: 8 inx12 in, Thickness: 0.04 in, Spark deflector for visor use without a safety cap, Fully dielectric, Visor secured with integral plastic lugs
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEADGEAR HDG20 FACE SHIELD 8X12X0.04 IN 76324-138 138-29076 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty