Would you like to log in again?
HEADGEAR 170SB BLACK BULK RATCHET BX40
Supplier: ORS NASCO, INC.
Headgear 170Sb Black Bulk Ratchet BX40^ Headgear, 170SB, Bulk, color: black, Suspension: Ratchet, Used With: Faceshield; JACKSON SAFETY* Brand Headgear, Spark deflector for visor use without a safety cap, Fully dielectric, Secured with integral plastic lugs, flying debris and splash protection
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEADGEAR 170SB BLACK BULK RATCHET BX40 76324-140 138-29077 Box of 40 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty