Would you like to log in again?
FACE SHIELD F50 PC 3465 8 INX15 1/2 IN
Supplier: ORS NASCO, INC.
Face Shield F50 Pc 3465 8 Inx15 1/2 In^ JACKSON SAFETY* Face Shield, F50, Model: 3465, Lens Material: Polycarbonate, Lens Coating/Shade: IR5, Dimension: 8 inx15 1/2in, Thickness: 0.06 in, Ideal for impact and heat resistance, 99.9% UV protection, Meets ANSI Z87.1 Standard
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F50 PC 3465 8 INX15 1/2 IN 76324-146 138-29080 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty